Learn More
Carson Optical™ Illuminated Handheld 5x Power 2.5 inch Aspheric LED Lighted Magnifier
SDPSCI_ED
Catalog No. S02019 Shop All Carson Optical Products
Change view
Click to view available options
Magnification:
5x
Catalog No. Magnification
S02019 5x
1 options
This item is not returnable. View return policy
Catalog No. S02019 Supplier Carson Optical™ Supplier No. SV50
Add to Cart
Add to Cart
This item is not returnable. View return policy
The Illuminated Handheld 5x Power Aspheric LED Lighted Magnifier is a 5x aspheric magnifier. Aspheric lens magnifiers allow for increased magnification and image clarity while greatly reducing edge to edge distortion.
5x aspheric magnifier that allows for increased magnification and image clarity while greatly reducing edge to edge distortion.
  • 2.5 in. crystal-clear acrylic lens allows it to be lightweight enough to use for everyday tasks.
  • The SV-50 is a great low vision aid and assists in reading small print.
  • Features an ultrabright LED light to brighten your viewing area.
  • Uses 3 AAA batteries (not included).
Specifications
Product Type Magnifier
Magnification 5x
Diameter (Englis) Lens 3.0 in.
Diameter (Metric) Lens 7.62 cm
Material Acrylic Lens