Learn More
Braintree Scientific Illuminated Restrainer
SDP
Catalog No. NC0220828
Encompass
Illuminated Rodent Tail Vein Unit offers "brite light" illumination system with rechargeable batteries and recharger unit. Features clear plastic tail insertion cone, formed white base housing batteries, on/off switch, and lights, base acts as battery charging unit, electrical plug for recharging batteries. **Sold for research purposes only and not for human use**
Catalog No. NC0220828 Supplier Braintree Scientific Supplier No. IRC562
Illuminated Rodent Tail Vein Unit offers "brite light" illumination system with rechargeable batteries and recharger unit. Features clear plastic tail insertion cone, formed white base housing batteries, on/off switch, and lights, base acts as battery charging unit, electrical plug for recharging batteries. **Sold for research purposes only and not for human use**