Learn More
Gold Biotechnology Inc Illumination™ Firefly Luciferase Enhanced Assay Kit 150 Assays
SDP
Catalog No. NC2012018
Encompass
Firefly Luciferase Assays are one of the leading reporter assays in the world in the measurement of gene function and gene regulation as well as being widely used in pharmaceutical screening. The luciferase assays are sensitive and convenient due to the absence of endogenous luciferase activity in most cell types and tissue. Firefly luciferase is a monomeric 62 kDa protein typically isolated from the firefly, Photinus pyralis, which catalyzes the ATP-dependent D-luciferin in the presence of oxygen and Mg2+ to oxyluciferin to produce a yellow to greenish light (~560 nm).
Catalog No. NC2012018 Supplier Gold Biotechnology Inc Supplier No. I930150
Firefly Luciferase Assays are one of the leading reporter assays in the world in the measurement of gene function and gene regulation as well as being widely used in pharmaceutical screening. The luciferase assays are sensitive and convenient due to the absence of endogenous luciferase activity in most cell types and tissue. Firefly luciferase is a monomeric 62 kDa protein typically isolated from the firefly, Photinus pyralis, which catalyzes the ATP-dependent D-luciferin in the presence of oxygen and Mg2+ to oxyluciferin to produce a yellow to greenish light (~560 nm).