Learn More
Cole-Parmer Cole-Parmer Microscopy Fiber Optic Illuminator, No Light Pipe; 115 VAC
Catalog No. 50628569 Shop All Cole-Parmer Products
Encompass_Preferred
These high-performance illuminators deliver 400,000 footcandles of high-intensity cold light. Minimal light leakage makes these illuminators ideal for your photosensitive applications. Audible noise and vibration is minimal, convenient side door makes changing lamps fast and easy, light pipes fit securely without need for adapters. Built-in handle stays out of the way.
Catalog No. 50-628-569 Supplier Cole-Parmer Supplier No. 4172300
These high-performance illuminators deliver 400,000 footcandles of high-intensity cold light. Minimal light leakage makes these illuminators ideal for your photosensitive applications. Audible noise and vibration is minimal, convenient side door makes changing lamps fast and easy, light pipes fit securely without need for adapters. Built-in handle stays out of the way.