Learn More
Electron Microscopy Sciences Illuminator
SDP
Catalog No. 5034426
Encompass_Preferred
This lighting device provides brightness around the base of the Macroscope-25. It can be moved vertically and positioned angularly to best highlight a specimen. It comes with batteries but it can also be operated with an optional AC adapter.
Catalog No. 50-344-26 Supplier Electron Microscopy Sciences Supplier No. 67974
This lighting device provides brightness around the base of the Macroscope-25. It can be moved vertically and positioned angularly to best highlight a specimen. It comes with batteries but it can also be operated with an optional AC adapter.