Learn More
Sigma Aldrich Fine Chemicals Biosciences illustraTM PuReTaq RTG PCR
Supplier:  Sigma Aldrich Fine Chemicals Biosciences GE27955701
Encompass
PuReTaq Ready-To-Go PCR Beads are pre-mixed predispensed single-dose reactions optimized for performing standard PCR amplifications. The use of recombinant PuReTaq DNA polymerase and other high-purity reagents ensures reliable and robust performance in both end point and real-time fluorescence-based PCR amplifications and ensures the lowest possible levels of contaminating prokaryotic and eukaryotic nucleic acids.The puReTaq Ready-To-Go PCR Beads are pre-formulated to ensure greater reproducibility between reactions minimize pipetting steps and reduce the potential for pipetting errors and contamination. The only additional reagents required are water primers and template DNA. The beads are provided pre-dispensed into either 0.2 ml or 0.5 ml PCR tubes. The 0.2 ml tubes are also supplied in a 96-well (8 x 12) plate format that allows individual strips of eight tubes to be easily removed. This flexibility allows use of either the entire 96-well plate strips of eight or individua
Catalog No. 50-290-4415