Learn More
TARGETMOL CHEMICALS INC ILOMASTAT 10MG
SDP
Catalog No. 502580614
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Ilomastat (GM6001) (GM6001 Galardin) is a broad spectrum matrix metalloprotease (MMP) inhibitor. purity: 99%
Catalog No. 50-258-0614 Supplier TARGETMOL CHEMICALS INC Supplier No. T274310MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Ilomastat (GM6001) (GM6001 Galardin) is a broad spectrum matrix metalloprotease (MMP) inhibitor. purity: 99%