Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ilomastat | 142880-36-2 | 388.5 g/mol | 10MM
SDP

Catalog No. NC2612413
Encompass_Preferred

Ilomastat | 142880-36-2 | 388.5 g/mol | 10MM

Catalog No. NC2612413 Supplier Medchemexpress LLC Supplier No. HY1576810MM1MLRECONST
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ilomastat | 142880-36-2 | 388.5 g/mol | 10MM

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.