Learn More
Selleck Chemical LLC Ilomastat (GM6001, Galardin) 5mg 142880-36-2
SDP
Supplier:  Selleck Chemical LLC S71575MG
Encompass_Preferred
Ilomastat (GM6001, Galardin) is a broad spectrum matrix metalloprotease (MMP) inhibitor for MMP-1, MMP-2, MMP-3, MMP-7, MMP-8, MMP-9, MMP-12, MMP-14, and MMP-26 with Ki of 0.4 nM, 0.5 nM, 27 nM, 3.7 nM, 0.1 nM, 0.2 nM, 3.6 nM, 13.4 nM, 0.36 nM, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-7983