Learn More
Cayman Chemical Iloperidone-d3, Purity: ≥99% (deuterated forms d1-d3), CAS: 1071167-49-1, 50 mg
SDP
Catalog No. 502910385
Encompass_Preferred
An internal standard for the quantification of iloperidone by GC- or LC-MS
Catalog No. 50-291-0385 Supplier Cayman Chemical Supplier No. 3519450 MG
An internal standard for the quantification of iloperidone by GC- or LC-MS