Learn More
Cayman Chemical Iloperidone metabolite P88, Purity: ≥98%, CAS: 133454-55-4, 10 mg
SDP
Catalog No. 502940303
Encompass_Preferred
An active metabolite of iloperidone; binds to 5-HT1B, 5-HT2A, and 5-HT2C (Kis = 5.1, 0.03, and 6.9 nM, respectively), α2b- and α2c-adrenergic (Kis = 6 and 1.3 nM, respectively), and dopamine D1, D2A, and D4 receptors (Kis = 9.5, 1.6, and 3.5 nM, respectively)
Catalog No. 50-294-0303 Supplier Cayman Chemical Supplier No. 2780610 MG
An active metabolite of iloperidone; binds to 5-HT1B, 5-HT2A, and 5-HT2C (Kis = 5.1, 0.03, and 6.9 nM, respectively), α2b- and α2c-adrenergic (Kis = 6 and 1.3 nM, respectively), and dopamine D1, D2A, and D4 receptors (Kis = 9.5, 1.6, and 3.5 nM, respectively)