Learn More
Cayman Chemical IloperIdon metabolIte P95 10mg
SDP
Supplier:  Cayman Chemical 2780710 MG
Encompass_Preferred
A metabolite of iloperidone; binds to 5-HT2A receptors and α1-, α2B- and α2C-adrenergic receptors (mean Kis = 7.08, 21.38, 83.18, and 47.86 nM, respectively); does not cross the blood-brain barrier
Catalog No. 50-293-8868