Learn More
Cayman Chemical Iloperidone metabolite P95, Purity: ≥98%, CAS: 475110-48-6, 5 mg
SDP
Catalog No. 502940019
Encompass_Preferred
A metabolite of iloperidone; binds to 5-HT2A receptors and α1-, α2B- and α2C-adrenergic receptors (mean Kis = 7.08, 21.38, 83.18, and 47.86 nM, respectively); does not cross the blood-brain barrier
Catalog No. 50-294-0019 Supplier Cayman Chemical Supplier No. 278075 MG
A metabolite of iloperidone; binds to 5-HT2A receptors and α1-, α2B- and α2C-adrenergic receptors (mean Kis = 7.08, 21.38, 83.18, and 47.86 nM, respectively); does not cross the blood-brain barrier