Learn More
Apexbio Technology LLC Iloperidone 133454-47-4 10mg
SDP
Catalog No. 502240346
Encompass_Preferred
Iloperidone (CAS 133454-47-4) is a small molecule antagonist of dopamine D2 and serotonin 5-HT2 receptors By inhibiting dopaminergic and serotonergic neurotransmission iloperidone modulates pathways implicated in psychotic disorders This compound is utilized in research to investigate receptor pharmacology and the underlying mechanisms of antipsychotic activity Its dual antagonism profile makes it relevant for studies examining the interplay between dopamine and serotonin signaling in neuropsychiatric disease models
This item is not returnable. View return policy
Catalog No. 50-224-0346 Supplier Apexbio Technology LLC Supplier No. A539910
Iloperidone (CAS 133454-47-4) is a small molecule antagonist of dopamine D2 and serotonin 5-HT2 receptors By inhibiting dopaminergic and serotonergic neurotransmission iloperidone modulates pathways implicated in psychotic disorders This compound is utilized in research to investigate receptor pharmacology and the underlying mechanisms of antipsychotic activity Its dual antagonism profile makes it relevant for studies examining the interplay between dopamine and serotonin signaling in neuropsychiatric disease models