Learn More
Cayman Chemical Iloprost-d4 25UG
SDP
Supplier:  Cayman Chemical 900036625 UG
Encompass_Preferred
An internal standard for the quantification of iloprost by GC- or LC-MS
Catalog No. 50-289-2444