Learn More
Cayman Chemical Iloprost-d4 50UG
SDP
Catalog No. 502893808
Encompass_Preferred
An internal standard for the quantification of iloprost by GC- or LC-MS
Catalog No. 50-289-3808 Supplier Cayman Chemical Supplier No. 900036650 UG
An internal standard for the quantification of iloprost by GC- or LC-MS