Learn More
Hamilton™ Microlab™ 600 Disposable Tip Hand Probe
Catalog No. 14818144 Shop All Hamilton Company Products
Change view
Click to view available options
Functional Range:
0.5 to 1000 μL
Catalog No. Functional Range
14-818-144 0.5 to 1000 μL
1 options
Catalog No. 14-818-144 Supplier Hamilton™ Supplier No. 6396002
Add to Cart
Add to Cart
Utilizes Hamilton's unique, environmentally-friendly ClickSure™ tips that have been validated for superior liquid handling.
  • When ClickSure tips are placed onto the probe, the O-ring on the probe compresses into the tip groove
  • Only requires a low insertion force to successfully load a tip
  • A more ergonomic experience for the user
  • Enables long-term use with minimal energy exerted
  • Probe accepts two ClickSure tips, 50μL and 1mL, which cover the entire pipetting volume range from 0.5 to 1000μL
Specifications
Disposable Single-use
For Use With (Equipment) Microlab 600 Advanced Diluter
Functional Range 0.5 to 1000 μL
Length (English) 5 ft. tube length
Length (Metric) 1549.4 mm tube length