Learn More
Selleck Chemical LLC IM-12 10mg
SDP
Supplier:  Selleck Chemical LLC S756610MG
Encompass_Preferred
IM-12 is a selective GSK-3? inhibitor with IC50 of 53 nM, and also enhances canonical Wnt signalling. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3286