Learn More
Apexbio Technology LLC IM-12 1129669-05-1 200mg
SDP
Catalog No. 5000087969
Encompass_Preferred
IM-12 (CAS 1129669-05-1) is a small-molecule inhibitor targeting glycogen synthase kinase-3 (GSK-3 ) displaying an IC50 of 53 nM GSK-3 is a serine/threonine kinase involved in neuronal development energy metabolism and Wnt/ -catenin signaling In ReNcell VM cells IM-12 inhibits GSK-3 resulting in increased -catenin accumulation enhanced TCF transcriptional activity and elevated neuronal differentiation markers The compound also extends cell doubling time and suppresses proliferation These properties make IM-12 valuable for investigating GSK-3 signaling in neurobiology cell proliferation and differentiation studies
Catalog No. 50-000-87969 Supplier Apexbio Technology LLC Supplier No. B4793200
Add to Cart
Add to Cart
IM-12 (CAS 1129669-05-1) is a small-molecule inhibitor targeting glycogen synthase kinase-3 (GSK-3 ) displaying an IC50 of 53 nM GSK-3 is a serine/threonine kinase involved in neuronal development energy metabolism and Wnt/ -catenin signaling In ReNcell VM cells IM-12 inhibits GSK-3 resulting in increased -catenin accumulation enhanced TCF transcriptional activity and elevated neuronal differentiation markers The compound also extends cell doubling time and suppresses proliferation These properties make IM-12 valuable for investigating GSK-3 signaling in neurobiology cell proliferation and differentiation studies