Learn More
Enzo Life Sciences IM-12 (25 mg). CAS: 1129669-05-1
SDP
Supplier:  Enzo Life Sciences BMLWN1020025
Encompass_Preferred
GSK-3β inhibitor (IC50=53nM (enzymatic), 3.8µM (cellular)). The compound significantly increases β-catenin levels, leads to TCF-induction and promotes neuronal differentiation in human neural progenitor cells via activation of canonical Wnt signaling. Alternative name: 3-(4-Fluorophenylethylamino)-1-methyl-4-(2-methyl-1H-indol-3-yl)-1H-pyrrole-2,5-dione. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (25 mg/ml) or in 100% ethanol (5 mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-2361