Learn More
Abcam IM 54 (2-(1H-Indol-3-yl)-3-pentylamino-maleimide), Necrosis inhibitor, 1MG
Catalog No. 502705377
Encompass_Preferred
MW 325.4 Da, Purity >98%. Potent, selective necrosis inhibitor (IC₅₀ = 3 μM, H₂O₂ exposed HeLa cells). Reduces oxidative stress-induced necrosis without effecting anticancer drug-induced apoptosis. Inhibits oxidative stress-induced necrotic cell death in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5377 Supplier Abcam Supplier No. AB1452311MG
MW 325.4 Da, Purity >98%. Potent, selective necrosis inhibitor (IC₅₀ = 3 μM, H₂O₂ exposed HeLa cells). Reduces oxidative stress-induced necrosis without effecting anticancer drug-induced apoptosis. Inhibits oxidative stress-induced necrotic cell death in vivo. .
The product is subject to the following: Abcam Restricted Use Statement