Learn More
Medchemexpress LLC HY-136093A 5mg Medchemexpress, IM156 CAS:1422365-94-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY136093A5MG
Encompass_Preferred
Medchemexpress, HY-136093A 5mg IM156 CAS:1422365-94-3 IM156 (HL156A; HL271 acetate), a chemical derivative of Metformin (HY-B0627), is a potent AMPK activator that increases AMPK phosphorylation. IM156 attenuates aging-associated cognitive impairment in animal model[2]. IM156 is a potent oxidative phosphorylation (OXPHOS) inhibitor which can be used for the research of solid tumors[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0394