Learn More
INDICAL INC IndiMag Pathogen IM48 Cartridge (6 x 8)
SDP
Catalog No. NC3371149
Encompass_Preferred
Prefilled reagent cartridges (32 well blocks) for automated magnetic-bead based extraction of nucleic acids on IndiMag 48/48s and IndiMag 2
Catalog No. NC3371149 Supplier INDICAL INC Supplier No. SP97654P608
Prefilled reagent cartridges (32 well blocks) for automated magnetic-bead based extraction of nucleic acids on IndiMag 48/48s and IndiMag 2