Learn More
Medchemexpress LLC IMAC Agarose (NTA) 6FF | 50 ML
SDP
Supplier:  Medchemexpress LLC HYK025550ML
Encompass_Preferred
MCE IMAC Agarose (NTA) 6FF is prepared by covalently coupling tetradentate nitrilotriacetic acid (NTA) to an agarose matrix. This chelating resin features high binding capacity, excellent specificity, and superior ligand stability, allowing flexible chelation with metal ions such as Zn2+, Ni2+, or Cu2+.
  • Suitable for purification of His-tagged recombinant proteins expressed in bacterial, mammalian, insect, and baculovirus systems.
  • Binding capacity depends on the type of metal ion used and specific experimental conditions, and should be evaluated by the user after metal ion loading.
  • Each 1 mL of total volume contains 0.5 mL of resin with a resin content of 50%.
Catalog No. 50-004-30194