Learn More
Biotage IMAC PHYTIP COLUMNS 1ML
SDP
Catalog No. NC2102051
Encompass
IMAC PhyTip columns (1 mL volume) containing 20 ul affinity media per column, Tecan. Incl. buffer kit
This item is not returnable. View return policy
Catalog No. NC2102051 Supplier Biotage Supplier No. PTT912003
IMAC PhyTip columns (1 mL volume) containing 20 ul affinity media per column, Tecan. Incl. buffer kit