Learn More
MSC Imada Pocket LED Stroboscope, 6 to 120,000 FPM, 0.007% Accuracy
Catalog No. 5000531556
Encompass
This stroboscope is designed for precise measurements, offering a broad operating range of 6 to 120,000 flashes per minute with an accuracy of up to 0.007%. It provides a resolution of 0.1 FPM for lower speeds and 1 FPM for higher speeds, making it suitable for various applications.
  • Pocket LED strobe type
  • NiMH battery chemistry for reliable power
  • LCD display for clear readings
  • Includes AC adapter/charger for convenience
  • Comes with a carrying case for portability
  • Batteries are included, ready for immediate use
Catalog No. 50-005-31556 Supplier MSC Supplier No. 67953471
This stroboscope is designed for precise measurements, offering a broad operating range of 6 to 120,000 flashes per minute with an accuracy of up to 0.007%. It provides a resolution of 0.1 FPM for lower speeds and 1 FPM for higher speeds, making it suitable for various applications.
  • Pocket LED strobe type
  • NiMH battery chemistry for reliable power
  • LCD display for clear readings
  • Includes AC adapter/charger for convenience
  • Comes with a carrying case for portability
  • Batteries are included, ready for immediate use