Learn More
Foxx Life Sciences Imafep™ MTO Fluorinated Ethylene Propylene (FEP) Supercritical Fluid Transfer Tubing 3mm (1/8") ID X 5mm (13/64") OD, 50 FT
Catalog No. 502061017
Encompass_Preferred
Imafep™ MTO Fluorinated Ethylene Propylene (FEP) Supercritical Fluid Transfer Tubing is designed for highly corrosive fluid transfer via piston pump or pressure transfer within the Pharmaceutical industry. Imafep™ is specially formulated to have transparent walls for visibility to meet global food and pharmaceutical grade standards. A smooth bore finish with low coefficient of friction ensures non-adherence of fluid particles. Steam sterilizable and resistant to most corrosive chemicals used in the Pharmaceutical industries, Imafep™ meets all Biocompatibility and Extractable requirements.• Manufactured in an ISO 9001, ISO 14001 and OHSAS 18001 certified facility• Excellent acid and alkali resistance• Steam, ETO, and Gamma Radiation sterilizable• Designed for Corrosive Solvent Transfer- Heptane, MDC, THF, Ethyl Acetate, CCL9• Service Temperature Range -60°C to +200°C.All standard dimensions are in mm and Inch dimensions are for reference only.
Catalog No. 50-206-1017 Supplier Foxx Life Sciences Supplier No. FEP00300050050
Imafep™ MTO Fluorinated Ethylene Propylene (FEP) Supercritical Fluid Transfer Tubing is designed for highly corrosive fluid transfer via piston pump or pressure transfer within the Pharmaceutical industry. Imafep™ is specially formulated to have transparent walls for visibility to meet global food and pharmaceutical grade standards. A smooth bore finish with low coefficient of friction ensures non-adherence of fluid particles. Steam sterilizable and resistant to most corrosive chemicals used in the Pharmaceutical industries, Imafep™ meets all Biocompatibility and Extractable requirements.• Manufactured in an ISO 9001, ISO 14001 and OHSAS 18001 certified facility• Excellent acid and alkali resistance• Steam, ETO, and Gamma Radiation sterilizable• Designed for Corrosive Solvent Transfer- Heptane, MDC, THF, Ethyl Acetate, CCL9• Service Temperature Range -60°C to +200°C.All standard dimensions are in mm and Inch dimensions are for reference only.