Learn More
Foxx Life Sciences EZLabpure™ Platinum-Cured Silicone Tubing, 6mm (15/64") ID X 9mm (23/64") OD, 25FT
Supplier:  Foxx Life Sciences FLO00600090025
Encompass_Preferred
EZLabpure™ Platinum-Cured Silicone Tubing is designed for general-purpose fluid transfer in food and pharmaceutical applications. Its smooth inner bore minimizes particle entrapment compared to peroxide-cured tubing, ensuring cleaner, more efficient flow. The tubing is developed to meet non-regulatory market requirements while maintaining high performance and reliability.
Features
Made from general-purpose platinum-grade silicone rubber
Made and packaged in an ISO Class 7 certified cleanroom
Clarity & Durability: Offers better transparency and longer shelf life than peroxide tubing
Safe Composition: Free from heavy metals and hazardous substances
Applications
Peristaltic pump-based fluid transfer
Chemical and analytical analysis
Sanitary application
Drug delivery and sampling
Food and beverages transfer
Certifications
NSF-51 Certification on Resin
FDA 21 CFR 177.2600
Phthalate/Bisphenol Free COA
TSE/BSE Certification
Catalog No. 50-206-1091