Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Image-iT™ Fixative Solutions

Catalog No. I28900
Encompass_Preferred
Change view
Click to view available options
Product Type:
Fixative
Formaldehyde
Paraformaldehyde
Quantity:
20 mL
2 x 50 mL
5 x 20 mL
10 x 10 mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Quantity
I28900 Fixative 10 x 10 mL
I28800 Paraformaldehyde 10 x 10 mL
I28700 Fixative 2 x 50 mL
FB002 Formaldehyde 20 mL
R37814 Formaldehyde 5 x 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. I28900 Supplier Invitrogen™ Supplier No. I28900
  • $91.50
    $75.03 / Each of 1

    Save $16.47

    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Image-iT Fixative Solutions are ready-to-use solutions provided in various formats to meet a variety of research needs. They are ideal for use with fluorescent proteins, such as GFP and RFP, and other small dye fluorophores. No more mixing of toxic fixative powders, just add to cell and fix.

Image-iT™ Fixative Solutions are ready-to-use solutions provided in various formats to meet expanding research needs.

Image-iT Fixative Solutions are ready-to-use fixative solutions provided in various formats to meet a variety of research needs. They are ideal for use with fluorescent proteins, such as GFP and RFP, and other small dye fluorophores. No more mixing of toxic fixative powders or dilution of concentrated solutions with non-optimal pH. Just add these fixative solutions to cells/tissue and incubate for 10-15 minutes at room temperature. They are ideal for use with any antibody-based detection method, including immunocytochemistry (ICC), immunofluorescence (IF), and immunohistochemistry (IHC).

Features of Image-iT Fixative Solutions include:

  • Ready-to-use
  • High-purity, imaging-grade fixatives with minimum background signal artifacts
  • Methanol-free
  • Optimized for secondary antibody labeling, fixed-cell dye staining, and colorimetric IHC

4% Formaldehyde or Paraformaldehyde Fixative Solutions

4% formaldehyde and paraformaldehyde are the most commonly used fixatives for immunocytochemistry (ICC), immunofluorescence (IF), and immunohistochemistry (IHC). Image-IT™ Paraformaldehyde 4% in 0.1M Phosphate Buffer pH 7.4 (Cat. No. I28800), is provided in ten vials pack with 10 mL each. Image-iT™ Formaldehyde 4% in PBS is provided in 20 mL size (Cat. No. FB002) or 5 x 20 mL size (Cat. No. R37814). Paraformaldehyde/formaldehyde 4% in 0.1 M phosphate buffer has ∼300 mOsm less osmolarity than in PBS, which helps maintain cellular morphology before the fixative has a chance to fix the inner cellular compartments. For paraformaldehyde/formaldehyde in PBS or phosphate buffer, just add the fixative to cells and incubate for 5-10 minutes at room temperature. Wash the cells in PBS after incubation and proceed to the blocking step.

Paraformaldehyde 3%, Glutaraldehyde 0.35% Fixative Solution

3% paraformaldehyde with 0.35% glutaraldehyde is a ready-to-use solution provided in methanol free, 0.1 M sodium cacodylate buffer. Yuan Qin, et al. (1) found that a mixture of paraformaldehyde and glutaraldehyde can preserve mitochondrial morphology better than either one alone.

Paraformaldehyde is small molecule that penetrates cells/tissue quickly, fixing proteins and other molecules in place. Glutaraldehyde is a bigger molecule with two reactive aldehyde groups that increase the potential for fixation and help create long chains of glutaraldehyde, generating a matrix. This ability helps preserve localization of proteins in complex environment such as mitochondria.

To fix cells/tissue, 5–10 minutes of incubation at room temperature is recommended. With glutaraldehyde, increased autofluorescence can be observed due to increased presence of reactive aldehyde groups. To help reduce this autofluorescence, approximately seven minutes of incubation with 0.1M glycine or freshly prepared 0.1% (w/v) sodium borohydride is recommended after fixation but before blocking.

Glyoxal 3% Fixative Solution

Glyoxal is a smaller molecule then formaldehyde or glutaraldehyde. It penetrates cells/tissue quickly, fixing the proteins, and preserving the cellular morphology better then formaldehyde (2). Glyoxal 3% Fixative Solution is a ready-to-use fixative solution provided in two separate bottles: Component A (92 mL) and Component B (8 mL), containing absolute Ethanol, Glyoxal Solution, Acetic Acid and water. Mix solution A and solution B in a empty vial, vortex the two solutions together and bring to a pH of 4.5 with the addition of 1M NaOH. Once mixed, this solution must be used immediately. To fix the cells/tissue, incubate cell/tissue at room temperature for approximately 10 minutes.

References

  1. Qin Y, Jiang W, Li A, Gao M, Liu H, Gao Y, Tian X, Gong G. The Combination of Paraformaldehyde and Glutaraldehyde Is a Potential Fixative for Mitochondria. Biomolecules. 2021 May 10;11(5):711.
  2. Richter KN, et al. Glyoxal as an alternative fixative to formaldehyde in immunostaining and super-resolution microscopy. EMBO J. 2018 Jan 4;37(1):139-159.
TRUSTED_SUSTAINABILITY

Specifications

Concentration PFA 3%, GA 0.35%
For Use With (Equipment) Microscope
Grade Fluorescence Grade
Product Line Image-iT
Product Type Fixative
Quantity 10 x 10 mL
Solution Type Paraformaldehyde
For Use With (Application) Immunocytochemistry (ICC), Immunohistochemistry (IHC), and Immunofluorescence (IF)
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.