Learn More
Motic™ Image-Pro v11.1 - Life Science Package for Motic (2D)
Catalog No. 01673119 Shop All Motic Group Products
Change view
Click to view available options
Color:
Purple
Catalog No. Color
01-673-119 Purple
1 options
Catalog No. 01-673-119 Supplier Motic™ Supplier No. MO7SW8
Add to Cart
Add to Cart
Request a Quote
This versatile solution combines advanced Machine Learning technology with a comprehensive suite of protocols to provide precise segmentation, classification, and automated measurements from 2D images.
Key Features:
  • Machine Learning-Powered Analysis: Leverage sophisticated Machine Learning algorithms for accurate segmentation and classification, ensuring automated and reliable measurements from 2D images.
  • Cell Biology Protocol Collection: Optimize your research with an extensive suite of protocols for analyzing complex cell biology-based measurements, including Cell Apoptosis, Cell Autophagy, Cell Confluence, Cell Count, Cell Morphology, Cell Proliferation, Cell Transfection, Lipid Droplet, Live/Dead Cell Viability, and Ring Region.
  • Versatility and Precision: Achieve high precision in your life science research across diverse applications with automated processes and detailed measurement capabilities.
  • This package also includes a 12-month subscription to a Standard Success Plan from Media Cybernetics, providing ongoing technical assistance and software onboarding so you can focus on the work that truly matters.
Specifications
Type Microscope Software
Autoclavable Not Autoclavable
Color Purple
Sterility Non-sterile