Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ IMAGEN™ Chlamydia Kit using Direct Immunofluorescence Assay

Catalog No. RK6101112 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RK6101112 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. RK6101112 Supplier Thermo Scientific™ Supplier No. K6101112
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rapidly detect and identify Chlamydiae in clinical specimens or cell cultures with this direct immunofluorescence assay.

The IMAGEN™ Chlamydia test is a qualitative direct immunofluorescence test for the detection of Chlamydia in human uro-genital and ophthalmic specimens and for the confirmation of Chlamydiae in cell culture.

Chlamydia trachomatis has been recognized as a frequent cause of sexually transmitted diseases1,2 and has been shown to cause a variety of human infections, some of which may be asymptomatic.

Use IMAGEN Chlamydia Direct Kit as a rapid, sensitive and specific method for direct detection and identification of Chlamydiae in clinical specimens or cell cultures.

  • Rapid results—within little over 30 minutes of receiving test sample or culture
  • Simple—ready-to-use reagents plus all IMAGEN assays follow the same basic methodology
  • Excellent sensitivity and specificity—high quality, well characterized antibodies used in the assay provide excellent sensitivity and specificity
  • Easy-to-read results
  • Long shelf life—typically 24 months

Chlamydiae are obligate intracellular parasites closely related to Gram-negative bacteria1. The genus Chlamydia contains three known species, Chlamydia trachomatis, Chlamydia psittaci and Chlamydia pneumoniae. The life cycle of Chlamydiae is complex but two main structures are recognized, the infectious elementary body and the reticulate body.

Currently, there are three main diagnostic methods used for the detection of Chlamydia in clinical specimens. Firstly, by isolation of the organism and visualization—viable elementary bodies present in clinical specimens infect tissue culture cells and the resultant inclusions are detected by staining techniques, e.g., iodine or immunofluorescence. Secondly, by direct demonstration of Chlamydia in clinical specimens using a fluorescein-labeled monoclonal antibody. Thirdly, by enzyme immunoassay (e.g., IDEIA™ PCE Chlamydia Kit) to determine the presence of Chlamydia antigen in clinical specimens3.

Direct immunofluorescence techniques utilising fluorescein-labeled monoclonal antibodies4,5,6 have been shown to have good correlation when compared to cell culture techniques for the detection of C. trachomatis.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific™ brand.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description IMAGEN™ Chlamydia Kit
Detectable Analytes Chlamydia Antigen
Type Chlamydia Testing Kit
For Use With (Application) Enzyme Immunoassay Detection
Includes 2 x positive control slide, mounting fluid, Chlamydia test reagent, instructions for use
Quantity 50 Tests
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.