Learn More
Thermo Scientific™ IMAGEN™ Herpes Simplex Virus (HSV) Kit using Direct Immunofluorescence Assay
Catalog No. RK6106112 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 Tests
Catalog No. Quantity
RK6106112 50 Tests
1 options
This item is not returnable. View return policy
Catalog No. RK6106112 Supplier Thermo Scientific™ Supplier No. K6106112
 Restricted Item
Add to Cart
Add to Cart
This item is not returnable. View return policy
Carry out rapid detection and identification of HSV1 and HSV2 in cell cultures with this direct immunofluorescence assay.
The IMAGEN™ Herpes Simplex Virus typing test is a qualitative direct immunofluorescence test for the detection and typing of HSV 1 and HSV 2 in cell cultures.
HSV is a common and universal infection of humans, associated with a wide range of clinical diseases in both immunocompetent and immunocompromised individuals. Both HSV 1 and 2 are frequently implicated in localised vesicular infections of the skin, conjunctiva and mucous membranes of the mouth or genitalia1, 2,3.
Use IMAGEN Herpes Simplex Virus (HSV) Kit as a rapid, sensitive and specific method for the detection and differentiation of HSV1 and HSV2 isolates in cell culture monolayers.
  • Rapid results- within little over 30 minutes of receiving test sample or culture
  • Simple- ready-to-use reagents plus all IMAGEN assays follow the same basic methodology
  • Excellent sensitivity and specificity- high quality, well-characterized antibodies used in the assay provide excellent sensitivity and specificity.
  • Easy-to-read results
  • Long Shelf Life- typically 24 months.
Herpes simplex virus is a DNA virus containing an icosahedral nucleocapsid surrounded by a lipid containing envelope. Human HSV is classified within the family Herpesviridae and is a member of the sub-family Alphaherpesvirinae. The genus Simplexvirus includes two type species of human herpes simplex virus, HSV 1 and HSV 24.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 2°C to 8°C
Description IMAGEN Herpes Simplex Virus (HSV) Kit
Detectable Analytes HSV 1/2
Includes 2 x 2 positive control slide, mounting fluid, HSV type 1 reagent, HSV type 2 reagent, instruction for use
Quantity 50 Tests