Learn More
Molecular Devices Molecular Devices Single-Cell Dispenser, Disposable Tip, 96/384 Well
Supplier:  Molecular Devices DISPENCELL
Encompass_Preferred
The DispenCell single-cell dispenser enables fast, gentle, and traceable isolation of individual cells into microplates, microwell arrays, or slides for downstream cloning and single-cell workflows. It combines a patented disposable sensing tip with single-cell analysis software to record electrical signals and provide immediate proof of clonality while maintaining high cell viability.
  • Fast, easy, and gentle single-cell isolation
  • Records electrical signals to provide immediate proof of clonality
  • Patented disposable sensing tip prevents cross-contamination
  • Compatible with 96-well and 384-well plates
  • No calibration or cleaning required
  • Sample preparation in about 10 minutes
  • Dispenses a 96-well plate in approximately 5 minutes
  • Operates at less than 0.1 psi to preserve viability
  • For research use only; not for diagnostic procedures
Catalog No. 50-000-14682
Only null left
Add to Cart
Request a Quote