Learn More
Molecular Devices Molecular Devices ImageXpress HCS.ai, High-Content Screening System, Spinning-disk Confocal, 8-Channel
Catalog No. 5000014735
Encompass_Preferred
The ImageXpress HCS.ai is an automated high-content imaging system that combines widefield and spinning-disk confocal fluorescence imaging with AI-driven analysis to enable high-throughput screening, live-cell assays, and 3D sample imaging.
  • Spinning-disk confocal and widefield imaging
  • Up to 8 fluorescent channels with laser option
  • Multiple confocal disk geometries including deep tissue option
  • High-throughput plate scanning with unattended runs
  • AI-driven image analysis workflows for complex phenotypes
Catalog No. 50-000-14735 Supplier Molecular Devices Supplier No. IXHCSAICF
Add to Cart
Add to Cart
Request a Quote
The ImageXpress HCS.ai is an automated high-content imaging system that combines widefield and spinning-disk confocal fluorescence imaging with AI-driven analysis to enable high-throughput screening, live-cell assays, and 3D sample imaging.
  • Spinning-disk confocal and widefield imaging
  • Up to 8 fluorescent channels with laser option
  • Multiple confocal disk geometries including deep tissue option
  • High-throughput plate scanning with unattended runs
  • AI-driven image analysis workflows for complex phenotypes