Learn More
Molecular Devices Molecular Devices ImageXpress HCS.ai, Widefield, sCMOS, 1-1536 Well
Supplier:  Molecular Devices IXHCSAIWF
Encompass_Preferred
The ImageXpress HCS.ai Widefield is an AI-enabled widefield high-content screening system for automated cellular imaging and phenotypic analysis. It combines high-speed sCMOS acquisition, flexible plate compatibility, and integrated AI-based analysis to accelerate 2D and 3D assay workflows and support live-cell and high-throughput screening applications.
  • High-throughput widefield imaging with sCMOS sensor.
  • AI-powered image analysis for rapid phenotypic profiling.
  • Supports microplates from 1 to 1536 wells and slide formats.
  • Optional environmental control for live-cell assays (temperature, CO2, O2).
  • Automated objective changer with 2X-60X magnifications.
Catalog No. 50-000-14700
Only null left
Add to Cart
Request a Quote