Learn More
Molecular Devices Molecular Devices ImageXpress Micro 4, Widefield Imager, 1-100x Objectives, sCMOS
Supplier:  Molecular Devices IXM4DB
Encompass_Preferred
The ImageXpress Micro 4 is an automated widefield high-content imaging system engineered for flexible, high-throughput cellular and whole-organism assays. It combines fast image acquisition, configurable optics, and comprehensive analysis software to support screening, live-cell studies, and 3D applications.
  • Widefield high-content imaging system.
  • Supports fluorescence, brightfield, phase contrast, colorimetric, and label-free imaging.
  • Upgradeable to confocal for volumetric 3D analysis.
  • Compatible with plates up to 1536-well and microscope slides.
  • sCMOS detector with >4 megapixels and 16-bit dynamic range.
  • User-changeable objectives, 1-100x range, 4-position objective changer.
  • Compatible with automation and integrated fluidics for liquid handling.
  • Optional environmental control for temperature, humidity, and CO2.
  • MetaXpress software for acquisition, analysis, and workflow automation.
Catalog No. 50-000-14697
Add to Cart
Request a Quote