Learn More
Electron Microscopy Sciences CoverWell™ Imaging Chamber 1 well 20 mm, 750 µl Volume
SDP
Catalog No. 5025146
Encompass_Preferred
CoverWell Imaging reusable press-to-seal silicone chambers form a removable enclosure for repeat staining or specimen repositioning. The chambers are designed to stabilize and support thick and free-floating specimens for confocal microscopy and imaging applications.
Diameter: 20 mm
Volume: 750 µl
O.D.: 25 mm x 25 mm
Catalog No. 50-251-46 Supplier Electron Microscopy Sciences Supplier No. 7032728
CoverWell Imaging reusable press-to-seal silicone chambers form a removable enclosure for repeat staining or specimen repositioning. The chambers are designed to stabilize and support thick and free-floating specimens for confocal microscopy and imaging applications.
Diameter: 20 mm
Volume: 750 µl
O.D.: 25 mm x 25 mm