Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences CoverWell™ Imaging Chamber 1 well 28 mm, 0.26mm depth, 200 µl Volume
SDP

Catalog No. 5025140
Encompass_Preferred

CoverWell Imaging reusable press-to-seal silicone chambers form a removable enclosure for repeat staining or specimen repositioning. The chambers are designed to stabilize and support thick and free-floating specimens for confocal microscopy and imaging applications.
Diameter: 28 mm
Depth: 0.26 mm
Volume: 200 µl
O.D.: 45 mm x 45 mm

Catalog No. 50-251-40 Supplier Electron Microscopy Sciences Supplier No. 7032704
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CoverWell Imaging reusable press-to-seal silicone chambers form a removable enclosure for repeat staining or specimen repositioning. The chambers are designed to stabilize and support thick and free-floating specimens for confocal microscopy and imaging applications.
Diameter: 28 mm
Depth: 0.26 mm
Volume: 200 µl
O.D.: 45 mm x 45 mm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.