Learn More
Molecular Devices Molecular Devices Microplate Reader, Absorbance, 190-1000 nm, 96/384-Well
Catalog No. 5000014386
Encompass_Preferred
The SpectraMax Plus 384 Microplate Reader is a UV/visible absorbance reader engineered for high-throughput spectrophotometric assays. It supports standard cuvettes, 96- and 384-well microplates, and offers monochromator-based wavelength selection across the UV-visible range, automated pathlength normalization for accurate cuvette-equivalent readings, and software integration for data acquisition and analysis.
  • UV/visible absorbance detection, 190-1000 nm.
  • Monochromator with 1 nm wavelength increments.
  • Compatible with 96-well and 384-well microplates and cuvettes.
  • Pathlength correction sensor for automatic 1 cm equivalent normalization.
  • Minimum volume 20 μL in 384-well plates.
  • Photometric range 0-4.000 OD.
  • Fast read speeds: 96-well 9 s/5 s, 384-well 29 s/16 s.
  • Temperature control from ambient +4 °C to 45 °C, uniformity ±0.5 °C at 37 °C.
Catalog No. 50-000-14386 Supplier Molecular Devices Supplier No. PLUS384
Add to Cart
Add to Cart
Request a Quote
The SpectraMax Plus 384 Microplate Reader is a UV/visible absorbance reader engineered for high-throughput spectrophotometric assays. It supports standard cuvettes, 96- and 384-well microplates, and offers monochromator-based wavelength selection across the UV-visible range, automated pathlength normalization for accurate cuvette-equivalent readings, and software integration for data acquisition and analysis.
  • UV/visible absorbance detection, 190-1000 nm.
  • Monochromator with 1 nm wavelength increments.
  • Compatible with 96-well and 384-well microplates and cuvettes.
  • Pathlength correction sensor for automatic 1 cm equivalent normalization.
  • Minimum volume 20 μL in 384-well plates.
  • Photometric range 0-4.000 OD.
  • Fast read speeds: 96-well 9 s/5 s, 384-well 29 s/16 s.
  • Temperature control from ambient +4 °C to 45 °C, uniformity ±0.5 °C at 37 °C.