Learn More
Imanis Life Sciences Imanis Life Sciences Cell Line, Human, B Lymphoblast, Fluc, EmGFP
Catalog No. NC3981232
Encompass
This human B-lymphoblast cell line, derived from Raji cells (ATCC® CCL-86™), expresses enhanced firefly luciferase (Fluc) and emerald green fluorescent protein (EmGFP). It is generated using a self-inactivating (SIN) lentiviral vector for enhanced biosafety.
  • Suitable for in vitro and in vivo studies.
  • Fluc enables noninvasive bioluminescent imaging of implanted cells.
  • EmGFP facilitates postmortem analysis by fluorescence microscopy or flow cytometry.
  • Mycoplasma free.
  • Grows in RPMI-1640 Medium with 10% FBS and 1% Penicillin/Streptomycin.
  • Maintain density between 5 x 105 and 2 x 106 cells/mL.
  • Characterized for morphology, reporter expression, and in vivo growth.
  • Biosafety level 2 reagents.
  • Store below -130°C in vapor phase above liquid nitrogen.
Catalog No. NC3981232 Supplier Imanis Life Sciences Supplier No. CL159
This human B-lymphoblast cell line, derived from Raji cells (ATCC® CCL-86™), expresses enhanced firefly luciferase (Fluc) and emerald green fluorescent protein (EmGFP). It is generated using a self-inactivating (SIN) lentiviral vector for enhanced biosafety.
  • Suitable for in vitro and in vivo studies.
  • Fluc enables noninvasive bioluminescent imaging of implanted cells.
  • EmGFP facilitates postmortem analysis by fluorescence microscopy or flow cytometry.
  • Mycoplasma free.
  • Grows in RPMI-1640 Medium with 10% FBS and 1% Penicillin/Streptomycin.
  • Maintain density between 5 x 105 and 2 x 106 cells/mL.
  • Characterized for morphology, reporter expression, and in vivo growth.
  • Biosafety level 2 reagents.
  • Store below -130°C in vapor phase above liquid nitrogen.