Learn More
Molecular Devices Molecular Devices Sterilant Kit, AquaMax Compatible, Concentrate, 2 Hour Soak
Catalog No. 5000014403
Encompass_Preferred
The Molecular Devices AquaMax Sterilant Kit is a two-part sterilant system designed to decontaminate and maintain AquaMax microplate washers. When prepared and applied as directed, it loosens protein and salt buildup, clears clogs, and removes endotoxin adherence from internal fluid paths.
  • Includes a concentrated sterilant and a separate diluent for on-instrument preparation.
  • Loosens protein and salt deposits and removes tough clogs from fluidic paths.
  • Reduces endotoxin adherence on interior fluid path surfaces.
  • Prepared by mixing the concentrate into the diluent, then transferring to a reservoir bottle.
  • Recommended two-hour soak after running the Clean program, followed by a distilled water flush.
  • Highly corrosive; use appropriate personal protective equipment and follow the supplied safety data.
Catalog No. 50-000-14403 Supplier Molecular Devices Supplier No. R8156
The Molecular Devices AquaMax Sterilant Kit is a two-part sterilant system designed to decontaminate and maintain AquaMax microplate washers. When prepared and applied as directed, it loosens protein and salt buildup, clears clogs, and removes endotoxin adherence from internal fluid paths.
  • Includes a concentrated sterilant and a separate diluent for on-instrument preparation.
  • Loosens protein and salt deposits and removes tough clogs from fluidic paths.
  • Reduces endotoxin adherence on interior fluid path surfaces.
  • Prepared by mixing the concentrate into the diluent, then transferring to a reservoir bottle.
  • Recommended two-hour soak after running the Clean program, followed by a distilled water flush.
  • Highly corrosive; use appropriate personal protective equipment and follow the supplied safety data.