Learn More
Molecular Devices Molecular Devices Membrane Potential Assay Kit, Evaluation, 96-Well
Catalog No. 5000014396
Encompass_Preferred
The FLIPR Membrane Potential Assay Kit (Evaluation) is a homogeneous, fluorescence-based reagent kit for detecting real-time changes in cellular membrane potential in microplate assays. It provides blue and red dye formulations and reagents sufficient for multiple 96-well assays.
  • Homogeneous fluorescence membrane potential assay for real-time measurements.
  • Includes blue and red component A dye vials and a buffer bottle.
  • Sufficient reagent for 10 microplates (96- or 384-well).
  • Designed for use with fluorescence plate readers and screening systems.
  • Suitable for ion channel screening and membrane potential functional assays.
  • Mix-and-read, no-wash protocol for high-throughput workflows.
Catalog No. 50-000-14396 Supplier Molecular Devices Supplier No. R8128
Add to Cart
Add to Cart
The FLIPR Membrane Potential Assay Kit (Evaluation) is a homogeneous, fluorescence-based reagent kit for detecting real-time changes in cellular membrane potential in microplate assays. It provides blue and red dye formulations and reagents sufficient for multiple 96-well assays.
  • Homogeneous fluorescence membrane potential assay for real-time measurements.
  • Includes blue and red component A dye vials and a buffer bottle.
  • Sufficient reagent for 10 microplates (96- or 384-well).
  • Designed for use with fluorescence plate readers and screening systems.
  • Suitable for ion channel screening and membrane potential functional assays.
  • Mix-and-read, no-wash protocol for high-throughput workflows.