Learn More
Molecular Devices Molecular Devices Phosphodiesterase Assay Kit, TR-FRET, FAM-cAMP, 800 data points
Catalog No. 5000014441
Encompass_Preferred
The IMAP TR-FRET phosphodiesterase evaluation assay kit is a homogeneous, mix-and-read biochemical assay for measuring PDE activity using IMAP progressive binding with time-resolved FRET detection. The kit supplies FAM-labeled cAMP and cGMP substrates with optimized reaction and binding buffers for high-throughput screening in 384-well format.
  • Provides FAM-labeled cAMP and FAM-labeled cGMP substrates.
  • Uses IMAP progressive binding and TR-FRET detection for homogeneous readouts.
  • Supplies progressive binding reagent, binding buffers, TR-FRET donor, and reaction buffers.
  • Configured for 20 μL reactions giving approximately 800 data points in 384-well format.
  • Includes reaction buffers with BSA and with Tween-20 for assay flexibility.
  • Store kit components at 4°C; aliquot substrates and store at -20°C; protect substrates from light.
  • Intended for in-vitro research use only; not for diagnostic procedures.
Catalog No. 50-000-14441 Supplier Molecular Devices Supplier No. R8176
The IMAP TR-FRET phosphodiesterase evaluation assay kit is a homogeneous, mix-and-read biochemical assay for measuring PDE activity using IMAP progressive binding with time-resolved FRET detection. The kit supplies FAM-labeled cAMP and cGMP substrates with optimized reaction and binding buffers for high-throughput screening in 384-well format.
  • Provides FAM-labeled cAMP and FAM-labeled cGMP substrates.
  • Uses IMAP progressive binding and TR-FRET detection for homogeneous readouts.
  • Supplies progressive binding reagent, binding buffers, TR-FRET donor, and reaction buffers.
  • Configured for 20 μL reactions giving approximately 800 data points in 384-well format.
  • Includes reaction buffers with BSA and with Tween-20 for assay flexibility.
  • Store kit components at 4°C; aliquot substrates and store at -20°C; protect substrates from light.
  • Intended for in-vitro research use only; not for diagnostic procedures.