Learn More
Molecular Devices Molecular Devices FLIPR Calcium Assay Kit, Explorer, 10-plate, 96/384/1536-well
Catalog No. 5000014402
Encompass_Preferred
The FLIPR Calcium 5 Assay Kit is a homogeneous fluorescent reagent kit designed to detect intracellular calcium flux in cell-based assays. It offers an enhanced signal window for improved sensitivity and is optimized for high-throughput screening on microplate readers and FLIPR systems.
  • Provides a large signal window for improved assay sensitivity.
  • Compatible with 96-, 384-, and 1536-well microplate formats.
  • Explorer kit supplies reagents sufficient for ten microplates.
  • Includes multiple reagent vials and assay/loading buffer for convenient setup.
  • Intended for research use only; not for diagnostic procedures.
  • Store components frozen on receipt; follow reconstitution and handling instructions for reagent stability.
Catalog No. 50-000-14402 Supplier Molecular Devices Supplier No. R8185
Add to Cart
Add to Cart
The FLIPR Calcium 5 Assay Kit is a homogeneous fluorescent reagent kit designed to detect intracellular calcium flux in cell-based assays. It offers an enhanced signal window for improved sensitivity and is optimized for high-throughput screening on microplate readers and FLIPR systems.
  • Provides a large signal window for improved assay sensitivity.
  • Compatible with 96-, 384-, and 1536-well microplate formats.
  • Explorer kit supplies reagents sufficient for ten microplates.
  • Includes multiple reagent vials and assay/loading buffer for convenient setup.
  • Intended for research use only; not for diagnostic procedures.
  • Store components frozen on receipt; follow reconstitution and handling instructions for reagent stability.