Learn More
Molecular Devices Molecular Devices Neurotransmitter Uptake Assay, Explorer, Homogeneous, 96/384-Well
Catalog No. 5000014504
Encompass_Preferred
Molecular Devices' Neurotransmitter Transporter Uptake Assay Explorer Kit provides a homogeneous, fluorescence-based method to measure dopamine, norepinephrine, and serotonin transporter (DAT, NET, SERT) uptake activity in live cells. The no-wash format supports kinetic and endpoint reads and is optimized for high-throughput screening and for use with transfected or endogenously expressing cells.
  • Homogeneous no-wash assay format.
  • Fluorescent substrate for DAT, NET, and SERT uptake.
  • Five lyophilized reagent vials, each sufficient for one 96- or 384-well plate.
  • Compatible with bottom-read fluorescence microplate readers and kinetic read modes.
  • Store reagents at -20°C; dye solution stable up to eight hours at room temperature or up to five days when aliquoted and frozen.
Catalog No. 50-000-14504 Supplier Molecular Devices Supplier No. R8173
Molecular Devices' Neurotransmitter Transporter Uptake Assay Explorer Kit provides a homogeneous, fluorescence-based method to measure dopamine, norepinephrine, and serotonin transporter (DAT, NET, SERT) uptake activity in live cells. The no-wash format supports kinetic and endpoint reads and is optimized for high-throughput screening and for use with transfected or endogenously expressing cells.
  • Homogeneous no-wash assay format.
  • Fluorescent substrate for DAT, NET, and SERT uptake.
  • Five lyophilized reagent vials, each sufficient for one 96- or 384-well plate.
  • Compatible with bottom-read fluorescence microplate readers and kinetic read modes.
  • Store reagents at -20°C; dye solution stable up to eight hours at room temperature or up to five days when aliquoted and frozen.