Learn More
Molecular Devices Molecular Devices IMAP TR-FRET Evaluation Kit, Homogeneous, 800 Data Points
Catalog No. 5000014488
Encompass_Preferred
The IMAP TR-FRET Evaluation Kit is a homogeneous, antibody-free TR-FRET assay system for evaluating kinase, phosphatase, and phosphodiesterase activity. The kit provides progressive binding reagents and TR-FRET donor reagents for mix-and-read assays and is sized for 800 data points in a 384-well format.
  • Homogeneous, mix-and-read TR-FRET assay format.
  • Antibody-free progressive binding chemistry to simplify assay design.
  • Reduced background through time-resolved detection for improved signal-to-noise.
  • Sized for 800 data points in a standard 384-well plate.
  • Includes binding reagents, TR-FRET donor, and reaction buffers for assay setup.
Catalog No. 50-000-14488 Supplier Molecular Devices Supplier No. R8161
The IMAP TR-FRET Evaluation Kit is a homogeneous, antibody-free TR-FRET assay system for evaluating kinase, phosphatase, and phosphodiesterase activity. The kit provides progressive binding reagents and TR-FRET donor reagents for mix-and-read assays and is sized for 800 data points in a 384-well format.
  • Homogeneous, mix-and-read TR-FRET assay format.
  • Antibody-free progressive binding chemistry to simplify assay design.
  • Reduced background through time-resolved detection for improved signal-to-noise.
  • Sized for 800 data points in a standard 384-well plate.
  • Includes binding reagents, TR-FRET donor, and reaction buffers for assay setup.