Learn More
Foxx Life Sciences Imapex™ MTO Peroxide Cured Silicone Transparent Tube 6.35mm(1/4") ID X 11.11mm(7/16") OD, 25 FT
Catalog No. 502061290
Encompass_Preferred
Imapex™ MTO Peroxide Grade silicone tubing is designed for non-critical fluid transfer, peristaltic pump use, and general engineering applications. Imapex™ is transparent and is made of high-quality peroxide silicone rubber tubing with excellent physical properties including tensile strength, flexibility, and resiliency.• Meets FDA 21 CFR 177.2600• Manufactured and Packaged in Dust Free environment• Odorless and Tasteless• Excellent flex fatigue resistance to enhance peristaltic pump life• Temperature resistance -80°C to + 230°C• BPA and Phthalate Free• TSE/BSE Free / ROHS compliantAll standard dimensions are in mm and Inch dimensions are for reference only.
Catalog No. 50-206-1290 Supplier Foxx Life Sciences Supplier No. PEX00630111025
Imapex™ MTO Peroxide Grade silicone tubing is designed for non-critical fluid transfer, peristaltic pump use, and general engineering applications. Imapex™ is transparent and is made of high-quality peroxide silicone rubber tubing with excellent physical properties including tensile strength, flexibility, and resiliency.• Meets FDA 21 CFR 177.2600• Manufactured and Packaged in Dust Free environment• Odorless and Tasteless• Excellent flex fatigue resistance to enhance peristaltic pump life• Temperature resistance -80°C to + 230°C• BPA and Phthalate Free• TSE/BSE Free / ROHS compliantAll standard dimensions are in mm and Inch dimensions are for reference only.