Learn More
Foxx Life Sciences EZLabpure™ Peroxide Cured Silicone Transparent Tubing, 4mm (5/32") ID X 8mm (5/16") OD, 50FT
GREENER_CHOICE
Catalog No. 502061246
Encompass_Preferred
EZLabpure™ Peroxide-Cured Silicone Tubing is designed for general, non-critical fluid transfer applications. This translucent tubing offers excellent flexibility and resilience, making it ideal for peristaltic pump use in routine non critical processes.
Features
Made from high-quality peroxide-cured silicone rubber
Manufactured and packaged in a dust-free facility
Odorless and tasteless for clean fluid transfer
Exhibits good compression set, tensile strength, and flex fatigue resistance to extend pump life
Wide Temperature Range: Operational from –80°C to +230°C
Applications
Peristaltic Pump
Liquid filling and transfer in Pharma & Biopharma Industries
Food and Beverages Industries
Certifications
Phthalates / Bis Phenol / Plasticizer free Certification
TSE/BSE Free Certification
5 years Shelf Life
FDA 21 CFR 177.2600 Certificate
Catalog No. 50-206-1246 Supplier Foxx Life Sciences Supplier No. PEX00400080050
EZLabpure™ Peroxide-Cured Silicone Tubing is designed for general, non-critical fluid transfer applications. This translucent tubing offers excellent flexibility and resilience, making it ideal for peristaltic pump use in routine non critical processes.
Features
Made from high-quality peroxide-cured silicone rubber
Manufactured and packaged in a dust-free facility
Odorless and tasteless for clean fluid transfer
Exhibits good compression set, tensile strength, and flex fatigue resistance to extend pump life
Wide Temperature Range: Operational from –80°C to +230°C
Applications
Peristaltic Pump
Liquid filling and transfer in Pharma & Biopharma Industries
Food and Beverages Industries
Certifications
Phthalates / Bis Phenol / Plasticizer free Certification
TSE/BSE Free Certification
5 years Shelf Life
FDA 21 CFR 177.2600 Certificate