Learn More
Medchemexpress LLC HY-101980 5mg , Imaradenant CAS:1321514-06-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1019805MG
Encompass_Preferred
HY-101980 5mg Imaradenant CAS: 1321514-06-0 AZD4635 (HTL1071) is a potent, selective and orally active adenosine A2A receptor (A2AR) antagonist. AZD4635 binds to human A2AR with a Ki of 1.7 nM and shows >30-fold selectivity over other adenosine receptors. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9430