Learn More
TARGETMOL CHEMICALS INC IMATINIB 1000MG
SDP
Catalog No. 502576565
Encompass_Preferred
Also available in 100 mg 200 mg 500 mg 5000 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Imatinib (STI571) is an inhibitor of the receptor tyrosine kinases c-Abl Bcr-Abl PDGFR and c-Kit (IC50 600/100/100 nM). purity: 99%
Catalog No. 50-257-6565 Supplier TARGETMOL CHEMICALS INC Supplier No. T62301G
Also available in 100 mg 200 mg 500 mg 5000 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Imatinib (STI571) is an inhibitor of the receptor tyrosine kinases c-Abl Bcr-Abl PDGFR and c-Kit (IC50 600/100/100 nM). purity: 99%